MOPS ≥99.5%, Ultrapure

Leverandør: Apollo Scientific
Warning

BIB2003-1KG BIB2003-250G BIB2003-500G
APOSBIB2003-1KGEA 2700 NOK
APOSBIB2003-1KG APOSBIB2003-250G APOSBIB2003-500G
MOPS ≥99.5%, Ultrapure
MOPS

Biological buffer - useful range 6.5-7.9


  • Heavy metals:<5 ppm
  • pKa @ 20°C: 7.20 + 0.2

Formel: C₇H₁₅NO₄S
Smeltepunkt: >277 °C
Flash punkt: 116 °C
Lagringstemperatur: Kjøleskap
MDL nummer: MFCD00006183
CAS-nummer: 1132-61-2
EINECS: 214-478-5

Bestill nå

SPESIFIKASJONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR