β-Nicotinamide adenine dinucleotide ≥97%

Leverandør: Apollo Scientific
Warning

Synonymer: Coenzyme I, β-NAD, Diphosphopyridine dinucleotide

BIB3011-1G BIB3011-5G
APOSBIB3011-1GEA 270 NOK
APOSBIB3011-1G APOSBIB3011-5G
β-Nicotinamide adenine dinucleotide ≥97%
β-Nicotinamide adenine dinucleotide
Formel: C₂₁H₂₇N₇O₁₄P₂
Lagringstemperatur: Fryseskap
MDL nummer: MFCD00036253
CAS-nummer: 53-84-9

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR