Nicotinamide adenine dinucleotide phosphate (reduced form) tetrasodium salt ≥98%

Leverandør: Apollo Scientific
Warning

BIB3014-100MG BIB3014-500MG
APOSBIB3014-100MGEA 750 NOK
APOSBIB3014-100MG APOSBIB3014-500MG
Nicotinamide adenine dinucleotide phosphate (reduced form) tetrasodium salt ≥98%
Nicotinamide adenine dinucleotide phosphate (reduced form) tetrasodium salt
Formel: C₂₁H₂₆N₇O₁₇P₃.4Na
Lagringstemperatur: Fryseskap
MDL nummer: MFCD00036263
CAS-nummer: 2646-71-1
EINECS: 220-163-3

Bestill nå

SPESIFIKASJONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR