5-Bromo-3-indolyl phosphate, p-toluidine salt

Leverandør: Apollo Scientific
Warning

Synonymer: Blue phosphate p-toluidine salt

BIB6014-1G BIB6014-500MG BIB6014-5G
APOSBIB6014-1GEA 1075 NOK
APOSBIB6014-1G APOSBIB6014-500MG APOSBIB6014-5G
5-Bromo-3-indolyl phosphate, p-toluidine salt
5-Bromo-3-indolyl phosphate, p-toluidine salt
Formel: C₈H₇BrNO₄P·C₇H₉N
Lagringstemperatur: Fryseskap
MDL nummer: MFCD03427361
CAS-nummer: 80008-69-1

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR