Spermidine phosphate salt hexahydrate

Leverandør: Apollo Scientific

BIB6271-25G BIB6271-5G BIB6271-10G
APOSBIB6271-25GEA 10275 NOK
APOSBIB6271-25G APOSBIB6271-5G APOSBIB6271-10G
Spermidine phosphate salt hexahydrate
Spermidine phosphate salt hexahydrate
Lagringstemperatur: Fryseskap
MDL nummer: MFCD04966492
CAS-nummer: 23273-82-7

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR