Cefazolin sodium salt

Leverandør: Apollo Scientific
Danger

BIC0101-1G BIC0101-5G
APOSBIC0101-1GEA 645 NOK
APOSBIC0101-1G APOSBIC0101-5G
Cefazolin sodium salt
Cefazolin sodium salt
Formel: C₁₄H₁₄N₈NaO₄S₃
Smeltepunkt: 170…172 °C (dec.)
Lagringstemperatur: Kjøleskap
MDL nummer: MFCD00056883
CAS-nummer: 27164-46-1
EINECS: 248-278-4

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR