Carbenicillin disodium salt

Leverandør: Apollo Scientific
Danger

BIC0109-25G BIC0109-5G BIC0109-100G
APOSBIC0109-25GEA 1975 NOK
APOSBIC0109-25G APOSBIC0109-5G APOSBIC0109-100G
Carbenicillin disodium salt
Carbenicillin disodium salt

Carbenicillin is an inhibitor of bacterial cell wall synthesisCell culture tested

Formel: C₁₇H₁₆N₂O₆S·2Na
Lagringstemperatur: Kjøleskap
MDL nummer: MFCD00077683
CAS-nummer: 4800-94-6
EINECS: 225-360-8

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR