Calcimycin (A23187)

Leverandør: Apollo Scientific
Warning

Synonymer: Calcium Ionophore A23187, Calcium Ionophore III, Antibiotic A23187

BIC1006-10MG BIC1006-5MG
APOSBIC1006-10MGEA 2900 NOK
APOSBIC1006-10MG APOSBIC1006-5MG
Calcimycin (A23187)
Calcimycin (A23187)

Calcium ionophore.

Formel: C₂₉H₃₇N₃O₆
MW: 523.63 g/mol
Smeltepunkt: 186 °C
Lagringstemperatur: Fryseskap
MDL nummer: MFCD00151202
CAS-nummer: 52665-69-7
EINECS: 258-084-1

Bestill nå

SPESIFIKASJONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR