Cytochalasin A

Leverandør: Apollo Scientific
Danger

BIC1013-10MG BIC1013-1MG BIC1013-5MG
APOSBIC1013-10MGEA 8925 NOK
APOSBIC1013-10MG APOSBIC1013-1MG APOSBIC1013-5MG
Cytochalasin A
Cytochalasin A

Oxidised analog of cytochalasin B (BIC1014).

Formel: C₂₉H₃₅NO₅
MW: 477.6 g/mol
Smeltepunkt: 193…195 °C
Lagringstemperatur: Fryseskap
MDL nummer: MFCD00005935
CAS-nummer: 14110-64-6
FN: 1544
ADR: 6.1 II

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR