Cytochalasin C

Leverandør: Apollo Scientific
Danger

BIC1015-1MG BIC1015-5MG
APOSBIC1015-1MGEA 2975 NOK
APOSBIC1015-1MG APOSBIC1015-5MG
Cytochalasin C
Cytochalasin C
Formel: C₃₀H₃₇NO₆
MW: 507.63 g/mol
Lagringstemperatur: Fryseskap
MDL nummer: MFCD00066080
CAS-nummer: 22144-76-9
FN: 1544
ADR: 6.1 II

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR