Cytochalasin D

Leverandør: Apollo Scientific
Danger

Synonymer: 7(S),18(R)-Dihydroxy-16(S),18-dimethyl-10-phenyl[11]cytochalasa-6(12),13(E),19(E)-triene-1,17-dione 21(R)-acetate

BIC1016-1MG BIC1016-5MG
APOSBIC1016-1MGEA 2700 NOK
APOSBIC1016-1MG APOSBIC1016-5MG
Cytochalasin D
Cytochalasin D
Formel: C₃₀H₃₇NO₆
MW: 507.63 g/mol
Smeltepunkt: 260…267 °C
Lagringstemperatur: Fryseskap
MDL nummer: MFCD00077706
CAS-nummer: 22144-77-0
EINECS: 244-804-1
FN: 1544
ADR: 6.1 II

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR