Cytochalasin E

Leverandør: Apollo Scientific
Danger

BIC1017-10MG BIC1017-1MG BIC1017-5MG
APOSBIC1017-10MGEA 8925 NOK
APOSBIC1017-10MG APOSBIC1017-1MG APOSBIC1017-5MG
Cytochalasin E
Cytochalasin E
Formel: C₂₈H₃₃NO₇
MW: 495.57 g/mol
Smeltepunkt: 206 °C
Lagringstemperatur: Fryseskap
CAS-nummer: 36011-19-5
EINECS: 252-835-7
FN: 1544
ADR: 6.1 II

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR