5-CFDA (5-Carboxyfluorescein diacetate)

Leverandør: Apollo Scientific

Synonymer: 5-Carboxy-di-O-acetylfluorescein, 5-CFDA, 5-Carboxyfluorescein diacetate

BIC1071-100MG
APOSBIC1071-100MGEA 2150 NOK
APOSBIC1071-100MG
5-CFDA (5-Carboxyfluorescein diacetate)
5-CFDA (5-Carboxyfluorescein diacetate)
Formel: C₂₅H₁₆O₉
Lagringstemperatur: Fryseskap
MDL nummer: MFCD00036872
CAS-nummer: 79955-27-4

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR