LC-SPDP (Succinimidyl 6-[3-(2-pyridylthio)propionamidohexanoate)

Leverandør: Apollo Scientific
Warning

Synonymer: LC SPDP

BICL208-100MG
APOSBICL208-100MGEA 4825 NOK
APOSBICL208-100MG
LC-SPDP (Succinimidyl 6-[3-(2-pyridylthio)propionamidohexanoate)
LC-SPDP (Succinimidyl 6-[3-(2-pyridylthio)propionamidohexanoate)

Spacer arm SPDPantibody-toxin conjugation


Ref: Carlsson,J.,et al (1978) Biochem.J.173, 723-737

Formel: C₁₈H₂₃N₃O₅S₂
MW: 425.522 g/mol
Smeltepunkt: 92…94 °C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00083163
CAS-nummer: 158913-22-5

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR