N-Succinimidyl Bromoacetate

Leverandør: Apollo Scientific
Warning

Synonymer: 2,5-Dioxopyrrolidin-1-yl 2-bromoacetate, N-(Bromoacetoxy)succinimide, Bromoacetic Acid N-Succinimidyl Ester, Bromoacetic acid N-hydroxysuccinimide ester

BICL212-1G
APOSBICL212-1GEA 1700 NOK
APOSBICL212-1G
N-Succinimidyl Bromoacetate
N-Succinimidyl Bromoacetate

Primary amine & Sulphhydryl reactiveProtein-Protein conjugation.

Formel: C₆H₆BrNO₄
MW: 236.02 g/mol
Lagringstemperatur: Fryseskap
MDL nummer: MFCD00058571
CAS-nummer: 42014-51-7

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR