rac-GR24

Leverandør: Apollo Scientific
Warning

Synonymer: (3aR*,8bS*,E)-3-(((R*)-4-methyl-5-oxo-2,5-dihydrofuran-2-yloxy)methylene)-3,3a,4,8b-tetrahydro-2H-indeno[1,2-b]furan-2-one

BICR114-100MG BICR114-20MG BICR114-5MG
APOSBICR114-100MGEA 53100 NOK
APOSBICR114-100MG APOSBICR114-20MG APOSBICR114-5MG
rac-GR24
rac-GR24
Formel: C₁₇H₁₄O₅
MW: 298.29 g/mol
Smeltepunkt: 156…157 °C
Tetthet: 1.364 g/cm³ (25 °C)
Lagringstemperatur: Fryseskap
MDL nummer: MFCD12405021
CAS-nummer: 76974-79-3

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR