(R)-allylglycinol hydrochloride

Leverandør: Apollo Scientific
Warning

Synonymer: (R)-2-aminopent-4-en-1-ol hydrochloride

BICR116-1G
APOSBICR116-1GEA 3875 NOK
APOSBICR116-1G
(R)-allylglycinol hydrochloride
(R)-allylglycinol hydrochloride
Formel: C₅H₁₁NO·ClH
MW: 137,608 g/mol
Lagringstemperatur: Fryseskap
MDL nummer: MFCD11052925
CAS-nummer: 926660-30-2

Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR