2'-Deoxycytidine

Leverandør: Apollo Scientific
Warning

BID1650-25G BID1650-50G BID1650-100G
APOSBID1650-25GEA 895 NOK
APOSBID1650-25G APOSBID1650-50G APOSBID1650-100G
2'-Deoxycytidine
2'-Deoxycytidine
Formel: C₉H₁₃N₃O₄
Lagringstemperatur: Fryseskap
MDL nummer: MFCD00006547
CAS-nummer: 951-77-9

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR