1,2-Naphthoquinone-4-sulphonic acid sodium salt

Leverandør: Apollo Scientific
Warning

Synonymer: Folin's reagent, Sodium 3,4-dioxo-3,4-dihydronaphthalene-1-sulphonate

BIF1006-25G BIF1006-5G
APOSBIF1006-25GEA 1300 NOK
APOSBIF1006-25G APOSBIF1006-5G
1,2-Naphthoquinone-4-sulphonic acid sodium salt
1,2-Naphthoquinone-4-sulphonic acid sodium salt
Formel: C₁₀H₅NaO₅S
MW: 260.20 g/mol
Smeltepunkt: 289 °C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00001700
CAS-nummer: 521-24-4
EINECS: 208-308-9

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR