Fluorescein isothiocyanate dextran

Leverandør: Apollo Scientific
Warning

Synonymer: FITC Dextran

BIF4025-100MG BIF4025-50MG
APOSBIF4025-100MGEA 2975 NOK
APOSBIF4025-100MG APOSBIF4025-50MG
Fluorescein isothiocyanate dextran
Fluorescein isothiocyanate dextran

Average molecular weight 70,000.

Lagringstemperatur: Kjøleskap
MDL nummer: MFCD00131092
CAS-nummer: 60842-46-8

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR