5-Aza-2'-deoxycytidine free base

Leverandør: Apollo Scientific
Warning

Synonymer: 5-Aza-2′-deoxycytidine, Decitabine, 5-Aza-1-(2-deoxy-beta-D-ribofuranosyl)cytosine, 4-Amino-1-((2R,4S,5R)-4-hydroxy-5-(hydroxymethyl)tetrahydrofuran-2-yl)-1,3,5-triazin-2(1H)-one, 2′-Deoxy-5-azacytidine 4-Amino-1-(2-deoxy-β-D-ribofuranosyl)-1,3,5-triazin-2(1H)-one Decitabine, 4-Amino-1-(2-deoxy-beta-D-ribofuranosyl)-1,3,5-triazin-2(1H)-one

BIFK0051-500MG BIFK0051-50MG BIFK0051-100MG
APOSBIFK0051-500MGEA 4925 NOK
APOSBIFK0051-500MG APOSBIFK0051-50MG APOSBIFK0051-100MG
5-Aza-2'-deoxycytidine free base
5-Aza-2'-deoxycytidine

  • Purity >99%

Formel: C₈H₁₂N₄O₄
Smeltepunkt: 193…203 °C
Lagringstemperatur: Fryseskap
MDL nummer: MFCD00043011
CAS-nummer: 2353-33-5

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR