Gentamicin sulphate

Leverandør: Apollo Scientific
Danger

Synonymer: NMD Inhibitor II, Garamycin, Gentamicin sulphate salt, Gentiomycin C

BIG0124-10G BIG0124-100G BIG0124-25G
APOSBIG0124-10GEA 305 NOK
APOSBIG0124-10G APOSBIG0124-100G APOSBIG0124-25G
Gentamicin sulphate
Gentamicin sulphate

An aminoglycoside antibiotic.


  • approx. 650 units/mg
  • Cell culture tested

Smeltepunkt: 217…237 °C
Lagringstemperatur: Kjøleskap
MDL nummer: MFCD00270181
CAS-nummer: 1405-41-0
EINECS: 215-778-9

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR