Ionomycin calcium salt

Leverandør: Apollo Scientific
Warning

BII0123-10MG BII0123-5MG
APOSBII0123-10MGEA 3500 NOK
APOSBII0123-10MG APOSBII0123-5MG
Ionomycin calcium salt
Ionomycin calcium salt

Potent and selective calcium ionophore.

Formel: C₄₁H₇₀CaO₉
Smeltepunkt: 204…206 °C
Lagringstemperatur: Fryseskap
MDL nummer: MFCD00151183
CAS-nummer: 56092-82-1

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR