Ionomycin free acid

Leverandør: Apollo Scientific
Warning

BII0421-10MG BII0421-5MG BII0421-25MG
APOSBII0421-10MGEA 4250 NOK
APOSBII0421-10MG APOSBII0421-5MG APOSBII0421-25MG
Ionomycin free acid
Ionomycin

Potent and selective calcium ionophore.

Formel: C₄₁H₇₂O₉
Smeltepunkt: 204…206 °C
Lagringstemperatur: Fryseskap
MDL nummer: MFCD06798385
CAS-nummer: 56092-81-0

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR