Imipenem monohydrate

Leverandør: Apollo Scientific
Warning

BII5001-100MG BII5001-10MG BII5001-1G
APOSBII5001-100MGEA 680 NOK
APOSBII5001-100MG APOSBII5001-10MG APOSBII5001-1G
Imipenem monohydrate
Imipenem monohydrate
Formel: C₁₂H₁₇N₃O₄S
Lagringstemperatur: Fryseskap
MDL nummer: MFCD09753321
CAS-nummer: 74431-23-5
EINECS: 264-734-5

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR