D(+)-Mannose 99,5%

Leverandør: Apollo Scientific
Warning

BIM0134-100G BIM0134-1KG BIM0134-500G
APOSBIM0134-100GEA 340 NOK
APOSBIM0134-100G APOSBIM0134-1KG APOSBIM0134-500G
D(+)-Mannose 99,5%
D(+)-Mannose

  • Heavy metals: <10ppm

Formel: C₆H₁₂O₆
Smeltepunkt: 133…140 °C
Tetthet: 1,54 g/cm³ (25 °C)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00064122
CAS-nummer: 3458-28-4
EINECS: 222-392-4

Bestill nå

SPESIFIKASJONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR