Salinomycin

Leverandør: Apollo Scientific
Danger

Synonymer: (R)-2-((2R,5S,6R)-6-((2S,3S,4S,6R)-6-((2S,5S,7R,9S,10S,12R,15R)-2-((2R,5R,6S)-5-Ethyl-5-hydroxy-6-methyltetrahydro-2H-pyran-2-yl)-15-hydroxy-2,10,12-trimethyl-1,6,8-trioxadispiro[4.1.57.35]pentadec-13-en-9-yl)-3-hydroxy-4-methyl-5-oxooctan-2-yl)-5-methyltetrahydro-2H-pyran-2-yl)butanoic acid

BIM0200-5MG BIM0200-10MG
APOSBIM0200-5MGEA 1500 NOK
APOSBIM0200-5MG APOSBIM0200-10MG
Salinomycin
Salinomycin
Formel: C₄₂H₇₀O₁₁
MW: 751.00 g/mol
Smeltepunkt: 112.5…113.5 °C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD25541652
CAS-nummer: 53003-10-4
FN: 3462
ADR: 6.1 II

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR