SATA (N-Succinimidyl S-Acetylthioglycolate)

Leverandør: Apollo Scientific
Warning

Synonymer: N-Succinimidyl-S-acetylthioacetate, S-Acetylthioglycolic Acid N-Succinimidyl Ester, SATA, 2,5-Dioxopyrrolidin-1-yl 2-(acetylthio)acetate

BIM101-100MG
APOSBIM101-100MGEA 1075 NOK
APOSBIM101-100MG
SATA (N-Succinimidyl S-Acetylthioglycolate)
SATA (N-Succinimidyl S-Acetylthioglycolate)

Primary amine reactive Introduces protected sulphhydryls.

Formel: C₈H₉NO₅S
MW: 231.22 g/mol
Smeltepunkt: 97 °C
Lagringstemperatur: Fryseskap
MDL nummer: MFCD00036891
CAS-nummer: 76931-93-6

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR