Sulphosuccinimidyl acetate (Sulpho-NHS-acetate)

Leverandør: Apollo Scientific
Warning

Synonymer: Sulpho-NHS-acetate, 1-(Acetyloxy)-2,5-dioxo-3-pyrrolidinesulphonic acid

BIM103-100MG
APOSBIM103-100MGEA 2250 NOK
APOSBIM103-100MG
Sulphosuccinimidyl acetate (Sulpho-NHS-acetate)
Sulphosuccinimidyl acetate (Sulpho-NHS-acetate)

Water soluble -Primary amine blocker.

Formel: C₄H₄NNaO₆S
Lagringstemperatur: Fryseskap
MDL nummer: MFCD01861959
CAS-nummer: 152305-87-8

Bestill nå

SPESIFIKASJONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR