4-Methylumbelliferyl-β-D-glucopyranoside

Leverandør: Apollo Scientific
Warning

Synonymer: 4-Methylumbelliferyl beta-D-Glucopyranoside, 4-Methylumbelliferyl β-D-Glucopyranoside, 4-MU-β-D-Glc, 4-Methyl-7-(((2S,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-2-yl)oxy)-2H-chromen-2-one, MUGlucosid

BIM1097-10G BIM1097-1G BIM1097-5G
APOSBIM1097-10GEA 3150 NOK
APOSBIM1097-10G APOSBIM1097-1G APOSBIM1097-5G
4-Methylumbelliferyl-β-D-glucopyranoside
4-Methylumbelliferyl-beta-D-glucopyranoside

Fluorescent substrate for b-D-galactosidase.

Formel: C₁₆H₁₈O₈
Smeltepunkt: 210…212 °C
Lagringstemperatur: Fryseskap
MDL nummer: MFCD00063694
CAS-nummer: 18997-57-4
EINECS: 242-736-7

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR