Okadaic acid free acid ≥98%

Leverandør: Apollo Scientific
Danger

BIM1576-1MG BIM1576-100UG BIM1576-50UG
APOSBIM1576-1MGEA 16425 NOK
APOSBIM1576-1MG APOSBIM1576-100UG APOSBIM1576-50UG
Okadaic acid free acid ≥98%
Okadaic acid

Potent inhibitor of protein phosphatases, especially the PP-1 and PP-2 classes, in numerous cell types Trends Biochem. Sci. 15: 98-102 (1990).

Formel: C₄₄H₆₈O₁₃
Smeltepunkt: 164…166 °C
Tetthet: 1,28 g/cm³ (25 °C)
Lagringstemperatur: Fryseskap
MDL nummer: MFCD00083455
CAS-nummer: 78111-17-8
FN: 3462
ADR: 6.1,I

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR