Zeaxanthin

Leverandør: Apollo Scientific

Synonymer: (1R,1'R)-4,4'-((1E,3E,5E,7E,9E,11E,13E,15E,17E)-3,7,12,16-tetramethyloctadeca-1,3,5,7,9,11,13,15,17-nonaene-1,18-diyl)bis(3,5,5-trimethylcyclohex-3-enol)

BIM1908-10MG BIM1908-25MG BIM1908-50MG
APOSBIM1908-10MGEA 1800 NOK
APOSBIM1908-10MG APOSBIM1908-25MG APOSBIM1908-50MG
Zeaxanthin
Zeaxanthin
Formel: C₄₀H₅₆O₂
MW: 568.89 g/mol
Smeltepunkt: 205…206 °C
Lagringstemperatur: Fryseskap
MDL nummer: MFCD08435940
CAS-nummer: 144-68-3

Bestill nå

SPESIFIKASJONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR