Phosphoramidon disodium salt
Leverandør: Apollo Scientific
BIMI0549-10MG BIMI0549-5MG
APOSBIMI0549-10MGEA 3500 NOK
APOSBIMI0549-10MG APOSBIMI0549-5MG
Phosphoramidon disodium salt
Proteiner
Inhibitor of thermolysin. Weak collagenase inhibitor. Does not inhibit chymotrypsin, papain, pepsin or trypsin.
Bestill nå
SPESIFIKASJONER
Protein/peptide name Phosphoramidon disodium salt
Environmentally Preferable
Protein synonyms N-(alpha-Rhamnopyranosyloxyhydroxyphosphinyl)-Leu-Trp disodium salt
Arter
Kilde