Nitrofurantoin

Leverandør: Apollo Scientific
Danger

BIN0136-100G BIN0136-250G BIN0136-500G
APOSBIN0136-100GEA 735 NOK
APOSBIN0136-100G APOSBIN0136-250G APOSBIN0136-500G
Nitrofurantoin
Nitrofurantoin

Nitrofurantoin is bactericidal in vitro to most gram-positive and gram-negative bacteria.

Formel: C₈H₆N₄O₅
Smeltepunkt: 267…269 °C (dec.)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00003224
CAS-nummer: 67-20-9
EINECS: 200-646-5

Bestill nå

SPESIFIKASJONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR