Penicillin V potassium salt

Leverandør: Apollo Scientific
Danger

BIP0143-25G BIP0143-5G
APOSBIP0143-25GEA 1450 NOK
APOSBIP0143-25G APOSBIP0143-5G
Penicillin V potassium salt
Penicillin V potassium salt

Penicillin V is an inhibitor of bacterial cell wall synthesisActive against gram-positive and some gram-negative bacteria. b-lactamase sensitive.


  • Potency: 1650mg/mg

Formel: C₁₆H₁₇KN₂O₅S
Smeltepunkt: 197…202 °C
Lagringstemperatur: Kjøleskap
MDL nummer: MFCD00051771
CAS-nummer: 132-98-9
EINECS: 205-086-5

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR