Banana powder
Leverandør: Apollo Scientific
BIPB852-100G
APOSBIPB852-100GEA 270 NOK
APOSBIPB852-100G
Banana powder
Engangsservise
A mixture of natural banana puree and maltodextrin. Use at a concentration of 30 to 50 g/L.
  • MFCD00130601