Coconut powder
Leverandør: Apollo Scientific
BIPC187-100G BIPC187-1KG BIPC187-500G
APOSBIPC187-100GEA 360 NOK
APOSBIPC187-100G APOSBIPC187-1KG APOSBIPC187-500G
Coconut powder
Engangsservise
A mixture of natural coconut puree and maltodextrin. Use at a concentration of 15 to 30 g/L.