Propynol ethoxylate

Leverandør: Apollo Scientific

BIPG1713-1G
APOSBIPG1713-1GEA 3100 NOK
APOSBIPG1713-1G
Propynol ethoxylate
Propynol ethoxylate
Formel: C₅H₈O₂
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00099294
CAS-nummer: 3973-18-0

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR