N-Phthalyl-L-tryptophan 98

Leverandør: Apollo Scientific
Warning

Synonymer: (S)-2-(1,3-Dioxoisoindolin-2-yl)-3-(1H-indol-3-yl)propanoic acid, RG 108, RG108

BIR1004-10MG BIR1004-25MG
APOSBIR1004-10MGEA 270 NOK
APOSBIR1004-10MG APOSBIR1004-25MG
N-Phthalyl-L-tryptophan 98
N-Phthalyl-L-tryptophan

DNA methyltransferase inhibitor. Reactivates tumour suppressor in gene expression in tumour cells.

Formel: C₁₉H₁₄N₂O₄
MW: 334.33 g/mol
Lagringstemperatur: Fryseskap
MDL nummer: MFCD08705332
CAS-nummer: 48208-26-0

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR