Spectinomycin dihydrochloride pentahydrate

Leverandør: Apollo Scientific
Warning

Synonymer: (2R,4aR,5aR,6S,7S,8R,9S,9aR,10aS)-4a,7,9-Trihydroxy-2-methyl-6,8-bis(methylamino)decahydro-4H-benzo[b]pyrano[2,3-e][1,4]dioxin-4-one dihydrochloride pentahydrate

BIS0147-25G BIS0147-5G
APOSBIS0147-25GEA 1075 NOK
APOSBIS0147-25G APOSBIS0147-5G
Spectinomycin dihydrochloride pentahydrate
Spectinomycin dihydrochloride pentahydrate

An aminocyclitol antibiotic which acts by binding to the 30S subunit of the bacterial ribosome and inhibiting protein synthesis.


  • Cell culture tested

Formel: C₁₄H₂₄N₂O₇·2HCl·5H₂O
Smeltepunkt: 210 °C
Lagringstemperatur: Kjøleskap
MDL nummer: MFCD00150886
CAS-nummer: 22189-32-8
EINECS: 244-554-3

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR