Streptomycin sulphate

Leverandør: Apollo Scientific
Warning

BIS0148-100G BIS0148-25G
APOSBIS0148-100GEA 750 NOK
APOSBIS0148-100G APOSBIS0148-25G
Streptomycin sulphate
Streptomycin sulphate

Streptomycin is an aminoglycoside antibiotic and has a bactericidal action against many gram-negative bacteria.


  • Cell culture tested

Formel: C₂₁H₃₉N₇O₁₂·1.5H₂SO₄
Lagringstemperatur: Kjøleskap
MDL nummer: MFCD00037023
CAS-nummer: 3810-74-0
EINECS: 223-286-0

Bestill nå

SPESIFIKASJONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR