Staurosporine (from Streptomyces species)

Leverandør: Apollo Scientific
Danger

BIS0504-100MG
APOSBIS0504-100MGEA 5900 NOK
APOSBIS0504-100MG
Staurosporine (from Streptomyces species)
Staurosporine

Potent inhibitor of phospholipid/calcium-dependent.protein kinase.

Formel: C₂₈H₂₆N₄O₃
Smeltepunkt: 270 °C
Lagringstemperatur: Fryseskap
MDL nummer: MFCD18252446
CAS-nummer: 62996-74-1

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR