Spermidine

Leverandør: Apollo Scientific
Danger

Synonymer: N1-(3-Aminopropyl)butane-1,4-diamine, N-(3-Aminopropyl)-1,4-diaminobutane, 4-Azaoctamethylenediamine

BIS5620-25G BIS5620-5G BIS5620-50G
APOSBIS5620-25GEA 3875 NOK
APOSBIS5620-25G APOSBIS5620-5G APOSBIS5620-50G
Spermidine
Spermidine

Used in the isolation of chromosomes.

Formel: C₇H₁₉N₃
MW: 145.25 g/mol
Kokepunkt: 246
Smeltepunkt: 24 °C
Tetthet: 0.925 g/cm³ (25 °C)
Flash punkt: 112 °C
Lagringstemperatur: Kjøleskap
MDL nummer: MFCD00008229
CAS-nummer: 124-20-9
FN: 2735
ADR: 8 II
Merck Indeks: 12,08893

Bestill nå

SPESIFIKASJONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR