AZD1480 99%

Leverandør: Apollo Scientific

Synonymer: (S)-5-Chloro-N2-(1-(5-fluoropyrimidin-2-yl)ethyl)-N4-(5-methyl-1H-pyrazol-3-yl)pyrimidine-2,4-diamine, AZD-1480

BISN0012-50MG
APOSBISN0012-50MGEA 2050 NOK
APOSBISN0012-50MG
AZD1480 99%
AZD1480

AZD1480 is a selective ATP-competitive JAK1/2 inhibitor with IC50 value of 1.3 and <0.4 nM, respectively, as well as shows inhibitory effect on JAK3 and Tyk2.

Formel: C₁₄H₁₄ClFN₈
MW: 348.77 g/mol
Lagringstemperatur: Fryseskap
MDL nummer: MFCD16038904
CAS-nummer: 935666-88-9

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR