LOXO-195 98%

Leverandør: Apollo Scientific

Synonymer: (3aR,10R)-5-Fluoro-1,2,3,3a,8,9,10,11-octahydro-10-methyl-12H-15,17-ethenopyrazolo[3,4-d]pyrido[2,3-k]pyrrolo[2,1-m][1,3,7]triazacyclotridecin-12-one, (13E,14E,22R,6R)-35-fluoro-6-methyl-7-aza-1(5,3)-pyrazolo[1,5-a]pyrimidina-3(3,2)-pyridina-2(1,2)-pyrrolidinacyclooctaphan-8-one, Selitrectinib

BISN0067-5MG
APOSBISN0067-5MGEA 840 NOK
APOSBISN0067-5MG
LOXO-195 98%
LOXO-195

LOXO-195 is a new-generation and selective TRK inhibitor.

Formel: C₂₀H₂₁FN₆O
MW: 380,43 g/mol
Lagringstemperatur: Kjøleskap
MDL nummer: MFCD31620755
CAS-nummer: 2097002-61-2

Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR