Filgotinib (GLPG0634)

Leverandør: Apollo Scientific

Synonymer: N-(5-(4-((1,1-Dioxidothiomorpholino)methyl)phenyl)-[1,2,4]triazolo[1,5-a]pyridin-2-yl)cyclopropanecarboxamide, Filgotinib

BISN0129-25MG
APOSBISN0129-25MGEA 825 NOK
APOSBISN0129-25MG
Filgotinib (GLPG0634)
Filgotinib (GLPG0634)

Filgotinib is a selective JAK12 inhibitor with IC50 values of 1028nM, with less potency against JAK3 and TYK2 with IC50 values of 810 nM and 116 nM, respectively.

Formel: C₂₁H₂₃N₅O₃S
MW: 425.5 g/mol
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD20527867
CAS-nummer: 1206161-97-8

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR