GDC0994

Leverandør: Apollo Scientific

Synonymer: RG-7842, Ravoxertinib, (S)-1-(1-(4-Chloro-3-fluorophenyl)-2-hydroxyethyl)-4-(2-((1-methyl-1H-pyrazol-5-yl)amino)pyrimidin-4-yl)pyridin-2(1H)-one, GDC0994, GDC-0994, RG 7842, GDC 0994, RG7842, 1-[(1S)-1-(4-chloro-3-fluorophenyl)-2-hydroxyethyl]-4-[2-[(2-methylpyrazol-3-yl)amino]pyrimidin-4-yl]pyridin-2-one

BISN0188-5MG
APOSBISN0188-5MGEA 825 NOK
APOSBISN0188-5MG
GDC0994
GDC0994

MAPK/ERK Signalling Pathway

Formel: C₂₁H₁₈ClFN₆O₂
MW: 440.86 g/mol
Lagringstemperatur: Fryseskap
MDL nummer: MFCD28143918
CAS-nummer: 1453848-26-4

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR