TAK-715

Leverandør: Apollo Scientific

Synonymer: N-(4-(2-Ethyl-4-(m-tolyl)thiazol-5-yl)pyridin-2-yl)benzamide

BISN0206-1MG
APOSBISN0206-1MGEA 415 NOK
APOSBISN0206-1MG
TAK-715
TAK-715

MAPK/ERK Signalling Pathway

Formel: C₂₄H₂₁N₃OS
MW: 399.5 g/mol
Lagringstemperatur: Kjøleskap
MDL nummer: MFCD17012805
CAS-nummer: 303162-79-0

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR