Raf1 Kinase Inhibitor I

Leverandør: Apollo Scientific

Synonymer: 5-Iodo-3-[(3,5-dibromo-4-hydroxyphenyl)methylene]-2-indolinone, 3-(3,5-dibromo-4-hydroxybenzylidene)-5-iodoindolin-2-one

BISN0227-1MG BISN0227-5MG BISN0227-10MG
APOSBISN0227-1MGEA 270 NOK
APOSBISN0227-1MG APOSBISN0227-5MG APOSBISN0227-10MG
Raf1 Kinase Inhibitor I
Raf1 Kinase Inhibitor I

MAPK/ERK Signalling Pathway

Formel: C₁₅H₈Br₂INO₂
MW: 520.9 g/mol
Lagringstemperatur: Romtemperatur
CAS-nummer: 220904-83-6

Bestill nå

SPESIFIKASJONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR