Anti-CD8 Mouse Antibody (Biotin)

Leverandør: Apollo Scientific
BITP2010-1MG
APOSBITP2010-1MGEA 0 NOK
APOSBITP2010-1MG
Anti-CD8 Mouse Antibody (Biotin)
Antistoffer
Bestill nå

SPESIFIKASJONER

Antigensymbol CD8
Environmentally Preferable
Konjugering Biotin
Reaktivitet Human
Vert Mouse
Isotype

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR